3-Ethyl-1-pentin-3-ol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Ethyl-3-hydroxy-1-pentin, 3-Ethylpent-1-in-3-ol
L06335.06
L06335.14
L06335.06EA
70.2
EUR
L06335.06
L06335.14
3-Ethyl-1-pentin-3-ol ≥98%
3-Ethyl-1-pentin-3-ol
Purity: 98%
CAS No.: 6285-06-9
Molecular Formula: C7H12O
Formula Weight: 112.17
Flash Point: 38°(100°F)
Refractive Index: 1.4380
Boiling Point: 138-140°
Beilstein Reference: 1740944
UN No.: 1987
MDL No.: MFCD00014392
Formel:
C₇H₁₂O Molecular Weight: 112.17 g/mol Siedepunkt: 138…140 °C Dichte: 0.873 Flammpunkt: 38 °C (100 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014392 CAS-Nummer: 6285-06-9 EINECS: 228-512-1 UN: 1987 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...