5-Methyl-1-hexin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
L06014.06
L06014.14
L06014.06EA
54.3
EUR
L06014.06
L06014.14
5-Methyl-1-hexin ≥98%
5-Methyl-1-hexin
Purity: 98%
CAS No.: 2203-80-7
Molecular Formula: (CH3)2CH(CH2)2C≡CH
Formula Weight: 96.17
Flash Point: -4°(24°F)
Refractive Index: 1.4075
Boiling Point: 91-93°
Beilstein Reference: 1731845
UN No.: 3295
MDL No.: MFCD00041610
Formel:
C₇H₁₂ Molecular Weight: 96,17 g/mol Siedepunkt: 91…93 °C Schmelzpunkt: –125 °C Dichte: 0,732 Flammpunkt: –4 °C (24 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041610 CAS-Nummer: 2203-80-7 UN: 3295 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...