3-Cyclopentylpropin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(2-Propinyl)cyclopentan, (2-Propynyl)cyclopentane
M-Clarity™ Qualitätsstufe = MQ100
632074-1G
632074-5G
SIAL632074-1GEA
74.2
EUR
SIAL632074-1G
SIAL632074-5G
3-Cyclopentylpropin, Sigma-Aldrich®
3-Cyclopentylpropin
Formel:
C₈H₁₂ Molecular Weight: 108.18 g/mol Siedepunkt: 132 Dichte: 0.827 g/cm³ (24.5 °C) |
MDL:
MFCD00041559 CAS-Nummer: 116279-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...