N-Chlorsuccinimid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: N-Chlor-1,4-butandisäureimid, N-Chlorbernsteinsäureimid, 1-Chlorbernsteinsäureimid, NCS, 1-Chlor-2,5-pyrrolidindion, Succinchlorimid, 1-Chlor-1,4-butandisäureimid, 1-Chlorazacyclopentan-2,5-dion

A10310.30 A10310.0B A10310.18
A10310.30EA 49.4 EUR
A10310.30 A10310.0B A10310.18
N-Chlorsuccinimid ≥98%
N-Chlorsuccinimid

Purity: 98% CAS No.: 128-09-6 Molecular Formula: C4H4ClNO2 Formula Weight: 133.53 Physical Form: Powder Density: 1.65 Appearance: White Odour: Chlorine-like Melting Point: 146-150° Beilstein Reference: 113915 Merck Reference: 14,2164 UN No.: 3261 MDL No.: MFCD00005511


Regioselective chlorination and oxidizing reagent

Formel: C₄H₄ClNO₂
Molecular Weight: 133,53 g/mol
Siedepunkt: 216,5 °C (1013 hPa)
Schmelzpunkt: 146…150 °C
Dichte: 1,65
Lagertemperatur: Raumtemperatur
MDL: MFCD00005511
CAS-Nummer: 128-09-6
EINECS: 204-878-8
UN: 3261
ADR: 8,II
Merck Index: 14,2164

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR