N-Hydroxysuccinimid ≥98%

Lieferant: Thermo Scientific

Synonyme: 1-Hydroxyazacyclopentan-2,5-dion, 1-Hydroxy-1,4-butandisäureimid, NHS, 1-Hydroxybernsteinsäureimid, 1-Hydroxy-2,5-pyrrolidindion, HOSu, N-Hydroxybernsteinsäureimid, N-Hydroxy-1,4-butandisäureimid

A10312.14 A10312.22 A10312.36
A10312.14EA 35.2 EUR
A10312.14 A10312.22 A10312.36
N-Hydroxysuccinimid ≥98%
N-Hydroxysuccinimid

Purity: 98+% CAS No.: 6066-82-6 Molecular Formula: C4H5NO3 Formula Weight: 115.09 Melting Point: 94-99° Beilstein Reference: 113913

Formel: C₄H₅NO₃
Molecular Weight: 115,09 g/mol
Siedepunkt: 363 °C (1013 hPa)
Schmelzpunkt: 94…99 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005516
CAS-Nummer: 6066-82-6
EINECS: 228-001-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR