N-Hydroxysuccinimid
Lieferant: BACHEM BIOCHEMICA
Synonyme:
1-Hydroxyazacyclopentan-2,5-dion, 1-Hydroxy-1,4-butandisäureimid, NHS, 1-Hydroxybernsteinsäureimid, 1-Hydroxy-2,5-pyrrolidindion, HOSu, N-Hydroxybernsteinsäureimid, N-Hydroxy-1,4-butandisäureimid
4025878.0100
4025878.0500
BACE4025878.0100EA
0
EUR
BACE4025878.0100
BACE4025878.0500
N-Hydroxysuccinimid
N-Hydroxysuccinimid
Formel:
C₄H₅NO₃ Molecular Weight: 115.09 g/mol Siedepunkt: 363 Schmelzpunkt: 93…95 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00005516 CAS-Nummer: 6066-82-6 EINECS: 228-001-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...