Pyrrolidin ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Tetrahydropyrrol, Tetramethylenimin

A14852.0F A14852.AE A14852.AP
A14852.0FEA 230 EUR
A14852.0F A14852.AE A14852.AP
Pyrrolidin ≥99%
Pyrrolidin

Purity: 99% CAS No.: 123-75-1 Molecular Formula: C4H9N Formula Weight: 71.12 Flash Point: 2°(35°F) Physical Form: Liquid Density: 0.856 Refractive Index: 1.4431 Sensitivity: Air Sensitive Appearance: Colourless to yellow Odour: Like rotten eggs Melting Point: -63° Boiling Point: 86-88° Solubility: Soluble in alcohol, ether, chloroform. Miscible with water Beilstein Reference: 102395 Merck Reference: 14,8015 UN No.: 1922 MDL No.: MFCD00005249

Formel: C₄H₉N
Molecular Weight: 71,12 g/mol
Siedepunkt: 86…88 °C
Schmelzpunkt: –63 °C
Dichte: 0,856
Flammpunkt: 2 °C (35 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005249
CAS-Nummer: 123-75-1
EINECS: 204-648-7
UN: 1922
ADR: 3,II
Merck Index: 14,8015

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR