1-Methylpiperidin ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: N-Methylpiperidine

L03398.09 L03398.22 L03398.36
L03398.09EA 25.4 EUR
L03398.09 L03398.22 L03398.36
1-Methylpiperidin ≥99%
1-Methylpiperidin

Purity: 99% CAS No.: 626-67-5 Molecular Formula: C6H13N Formula Weight: 99.18 Flash Point: 3°(37°F) Refractive Index: 1.4378 Odour: Amine-like pH: 12 Melting Point: -50° Boiling Point: 105-107° Beilstein Reference: 1073 UN No.: 2399 MDL No.: MFCD00006491

Formel: C₆H₁₃N
Molecular Weight: 99,18 g/mol
Siedepunkt: 105…107 °C
Schmelzpunkt: –50 °C
Dichte: 0,816
Flammpunkt: 3 °C (37 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006491
CAS-Nummer: 626-67-5
EINECS: 210-959-9
UN: 2399
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR