2-Methylpiperidin ≥98%

Lieferant: Thermo Scientific
Gefahr

L02567.AE L02567.AP
L02567.AEEA 40.5 EUR
L02567.AE L02567.AP
2-Methylpiperidin ≥98%
2-Methylpiperidin

Purity: 98+% CAS No.: 109-05-7 Molecular Formula: C6H13N Formula Weight: 99.18 Flash Point: 8°(46°F) Refractive Index: 1.4470 Odour: Amine-like Melting Point: -4° Boiling Point: 118-119° Beilstein Reference: 79804 UN No.: 2733 MDL No.: MFCD00005982

Formel: C₆H₁₃N
Molecular Weight: 99,18 g/mol
Siedepunkt: 118…119 °C
Schmelzpunkt: –4 °C
Dichte: 0,842
Flammpunkt: 8 °C (46 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005982
CAS-Nummer: 109-05-7
EINECS: 203-642-1
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR