2-Methylpyrrolidin, Sigma-Aldrich®

Lieferant: Merck

478059-10G 478059-2G
SIAL478059-10GEA 503 EUR
SIAL478059-10G SIAL478059-2G
2-Methylpyrrolidin, Sigma-Aldrich®
2-Methylpyrrolidin
Formel: C₅H₁₁N
Molecular Weight: 85,15 g/mol
MDL: MFCD00014491
CAS-Nummer: 765-38-8
EINECS: 212-144-3
UN: 2924
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR