3-Methylpiperidin ≥98%

Lieferant: Thermo Scientific
Gefahr

L13408.14 L13408.22
L13408.14EA 25.5 EUR
L13408.14 L13408.22
3-Methylpiperidin ≥98%
3-Methylpiperidin

Purity: 98% CAS No.: 626-56-2 Molecular Formula: C6H13N Formula Weight: 99.18 Flash Point: 8°(46°F) Refractive Index: 1.4470 Odour: Amine-like Melting Point: -24° Boiling Point: 124-125° Beilstein Reference: 79807 UN No.: 2733 MDL No.: MFCD00005994

Formel: C₆H₁₃N
Molecular Weight: 99,18 g/mol
Siedepunkt: 124…125 °C
Schmelzpunkt: –24 °C
Dichte: 0,845
Flammpunkt: 8 °C (46 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005994
CAS-Nummer: 626-56-2
EINECS: 210-953-6
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR