Morpholin ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

A10355.22 A10355.0E A10355.36
A10355.22EA 24.3 EUR
A10355.22 A10355.0E A10355.36
Morpholin ≥99%
Morpholin

Purity: 99% CAS No.: 110-91-8 Molecular Formula: C4H9NO Formula Weight: 87.12 Flash Point: 35°(95°F) Physical Form: Liquid Density: 1.007 Refractive Index: 1.4540 Sensitivity: Hygroscopic Appearance: Colourless Odour: Amine-like Melting Point: -5° Boiling Point: 129° Beilstein Reference: 102549 Merck Reference: 14,6277 UN No.: 2054 MDL No.: MFCD00005972

Formel: C₄H₉NO
Molecular Weight: 87.12 g/mol
Siedepunkt: 129 °C
Schmelzpunkt: –5 °C
Dichte: 1.007
Flammpunkt: 35 °C (95 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005972
CAS-Nummer: 110-91-8
EINECS: 203-815-1
UN: 2054
ADR: 8 I
Merck Index: 12,06362
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9fb9ce2adfd770be2ae8d5f63ea6220bef0f781&vid=6b2bfa885631c3f4e0681813373d468dcc171b83&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR