2-Methylbenzimidazol 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Methyl-1H-benzimidazol
126330050
126331000
ACRO126330050EA
28
EUR
ACRO126330050
ACRO126331000
2-Methylbenzimidazol 98%
2-Methylbenzimidazol
Formel:
C₈H₈N₂ Molecular Weight: 132.16 g/mol Siedepunkt: 339 Schmelzpunkt: 175…179 °C |
MDL:
MFCD00005598 CAS-Nummer: 615-15-6 EINECS: 210-411-9 |
Spezifikation Testergebnisse
Appearance | White to light yellow to beige Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 175 to 179 °C |
HPLC | ≥97.5 % |
Residue after evaporation | ≤0.001 % |
Acidity | ≤0.0002 meq/g |
Substance darkened by H2SO4 | Passes test |
Iron (Fe) | ≤1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...