Benz[a]anthracen 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Benz(alpha)anthracen, BaA, Benzo(alpha)anthracen, 1,2-Benzanthracen

105250010 105252500
ACRO105250010EA 110 EUR
ACRO105250010 ACRO105252500
Benz[a]anthracen 99%
Benz[a]anthracen
Formel: C₁₈H₁₂
Molecular Weight: 228,29 g/mol
Siedepunkt: 437,6 °C (760 mmHg)
Schmelzpunkt: 158…161 °C
Dichte: 1,245 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003599
CAS-Nummer: 56-55-3
EINECS: 200-280-6
UN: 3077
ADR: 9,III
Merck Index: 12,01089

Bestellen

Spezifikation Testergebnisse

Appearance Beige to yellow-green Crystalline powder
Infrared spectrum Conforms
Melting point 158 to 161 °C
GC ≥98.5 %
Optimum lasing 518 nm (in Methanol)
Suitability (for use as a laser dye) Passes test
Molar absorption coefficient 19900 l/(mol.cm) (at Lambda max)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR