4,7-Dichlorisatin ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4,7-Dichlor-2,3-indolindion, 4,7-Dichlor-1H-indol-2,3-dion, 4,7-Dichlor-2,3-dioxoindolin, 4,7-Dichlorindolin-2,3-dion

A14853.03 A14853.06
A14853.03EA 65.3 EUR
A14853.03 A14853.06
4,7-Dichlorisatin ≥98%
4,7-Dichlorisatin

Purity: 98% CAS No.: 18711-13-2 Molecular Formula: C8H3Cl2NO2 Formula Weight: 216.02 Melting Point: 246-249° Beilstein Reference: 168731

Formel: C₈H₃Cl₂NO₂
Molecular Weight: 216,02 g/mol
Schmelzpunkt: 246…249 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00047214
CAS-Nummer: 18711-13-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR