4,7-Dichlorisatin
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4,7-Dichlor-2,3-indolindion, 4,7-Dichlor-1H-indol-2,3-dion, 4,7-Dichlor-2,3-dioxoindolin, 4,7-Dichlorindolin-2,3-dion
S13100.1
S13100.10
MAYBS13100.1EA
0
EUR
MAYBS13100.1
MAYBS13100.10
4,7-Dichlorisatin
4,7-Dichlorisatin
Formel:
C₈H₃Cl₂NO₂ Molecular Weight: 216.02 g/mol |
MDL:
MFCD00047214 CAS-Nummer: 18711-13-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...