5-Chlorisatin ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 5-Chlor-2,3-indolindion, 5-Chlorindolin-2,3-dion, 5-Chlor-1H-indol-2,3-dion, 5-Chlor-2,3-dioxoindolin

A13125.06 A13125.14 A13125.22
A13125.06EA 20.9 EUR
A13125.06 A13125.14 A13125.22
5-Chlorisatin ≥98%
5-Chlorisatin

Purity: 98% CAS No.: 17630-76-1 Molecular Formula: C8H4ClNO2 Formula Weight: 181.58 Melting Point: ca 247° dec. Beilstein Reference: 383759

Formel: C₈H₄ClNO₂
Siedepunkt: 361 °C (1013 hPa)
Schmelzpunkt: ∼ 247 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00014567
CAS-Nummer: 17630-76-1
EINECS: 241-614-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR