5-Chlorisatin
Lieferant: FLUOROCHEM
Synonyme:
5-Chlor-2,3-indolindion, 5-Chlorindolin-2,3-dion, 5-Chlor-1H-indol-2,3-dion, 5-Chlor-2,3-dioxoindolin
001745-25G
001745-100G
FLUO001745-25GEA
86
EUR
FLUO001745-25G
FLUO001745-100G
5-Chlorisatin
5-Chlorisatin
Formel:
C₈H₄ClNO₂ Siedepunkt: 361 °C (1013 hPa) Schmelzpunkt: 247 °C |
MDL:
MFCD00014567 CAS-Nummer: 17630-76-1 EINECS: 241-614-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...