5-Fluorisatin 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Fluor-1H-indol-2,3-dion, 5-Fluorindolin-2,3-dion, 5-Fluor-2,3-indolindion, 5-Fluor-2,3-dioxoindolin
367700050
367700250
ACRO367700050EA
64.6
EUR
ACRO367700050
ACRO367700250
5-Fluorisatin 98%
5-Fluorisatin
Formel:
C₈H₄FNO₂ Molecular Weight: 165,12 g/mol Schmelzpunkt: 224…228 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00022795 CAS-Nummer: 443-69-6 |
Spezifikation Testergebnisse
Appearance | Red to brown Powder |
Infrared spectrum | Conforms |
Melting point | 224 to 230 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...