5-Fluorisatin ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Fluor-1H-indol-2,3-dion, 5-Fluorindolin-2,3-dion, 5-Fluor-2,3-indolindion, 5-Fluor-2,3-dioxoindolin

B20831.03 B20831.06 B20831.14
B20831.03EA 22.6 EUR
B20831.03 B20831.06 B20831.14
5-Fluorisatin ≥98%
5-Fluorisatin

Purity: 98% CAS No.: 443-69-6 Molecular Formula: C8H4FNO2 Formula Weight: 165.12 Melting Point: 224-227° Beilstein Reference: 131241

Formel: C₈H₄FNO₂
Molecular Weight: 165,12 g/mol
Schmelzpunkt: 224…227 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00022795
CAS-Nummer: 443-69-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR