1-Methylisatin

Lieferant: FLUOROCHEM

Synonyme: 1-Methylindolin-2,3-dion, 1-Methyl-1H-indol-2,3-dion, N-Methyl-2,3-dioxoindolin, N-Methyl-2,3-dihydroindol-2,3-dion, N-Methyl-1H-indol-2,3-dion, N-Methylindolin-2,3-dion, 1-Methyl-2,3-indolindion, N-Methylisatin, 1-Methyl-2,3-dioxoindolin, N-Methyl-2,3-indolindion, 1-Methyl-2,3-dihydroindol-2,3-dion

011197-25G 011197-10G
FLUO011197-25GEA 243 EUR
FLUO011197-25G FLUO011197-10G
1-Methylisatin
1-Methylisatin
Formel: C₉H₇NO₂
Molecular Weight: 161,16 g/mol
Schmelzpunkt: 139 °C
Dichte: 1,4 g/cm³ (20 °C)
MDL: MFCD00005812
CAS-Nummer: 2058-74-4
EINECS: 218-164-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR