1-Phenylisatin ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: N-Phenyl-2,3-indolindion, 1-Phenyl-1H-indol-2,3-dion, N-Phenyl-2,3-dioxoindolin, 1-Phenyl-2,3-dioxoindolin, 1-Phenyl-2,3-dihydroindol-2,3-dion, 1-Phenylindolin-2,3-dion, N-Phenyl-1H-indol-2,3-dion, N-Phenylindolin-2,3-dion, N-Phenylisatin, N-Phenyl-2,3-dihydroindol-2,3-dion, 1-Phenyl-2,3-indolindion

L11388.03 L11388.06
L11388.03EA 48.9 EUR
L11388.03 L11388.06
1-Phenylisatin ≥98%
1-Phenylisatin

Purity: 98% CAS No.: 723-89-7 Molecular Formula: C14H9NO2 Formula Weight: 223.23 Melting Point: 139-142° Beilstein Reference: 164531

Formel: C₁₄H₉NO₂
Molecular Weight: 223,23 g/mol
Siedepunkt: 404 °C (1013 hPa)
Schmelzpunkt: 139…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00082681
CAS-Nummer: 723-89-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR