1-Phenylisatin
Lieferant: BIONET RESEARCH
|
Gefahr
|
Synonyme:
N-Phenyl-2,3-indolindion, 1-Phenyl-1H-indol-2,3-dion, N-Phenyl-2,3-dioxoindolin, 1-Phenyl-2,3-dioxoindolin, 1-Phenyl-2,3-dihydroindol-2,3-dion, 1-Phenylindolin-2,3-dion, N-Phenyl-1H-indol-2,3-dion, N-Phenylindolin-2,3-dion, N-Phenylisatin, N-Phenyl-2,3-dihydroindol-2,3-dion, 1-Phenyl-2,3-indolindion
6L-325S.500
6L-325S.1
6L-325S.5
6L-325S.10
BIOT6L-325S.500EA
0
EUR
BIOT6L-325S.500
BIOT6L-325S.1
BIOT6L-325S.5
BIOT6L-325S.10
1-Phenylisatin
1-Phenylisatin
Formel:
C₁₄H₉NO₂ Molecular Weight: 223,23 g/mol Siedepunkt: 404 °C (1013 hPa) Schmelzpunkt: 197…198 °C |
MDL:
MFCD00082681 CAS-Nummer: 723-89-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...