Dichlon 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2,3-Dichlor-1,4-naphthochinon, 2,3-Chlorbenzo(e)cyclohex-2-en-1,4-dion
113480250
113481000
113482500
ACRO113480250EA
22.2
EUR
ACRO113480250
ACRO113481000
ACRO113482500
Dichlon 98%
Dichlon
Formel:
C₁₀H₄Cl₂O₂ Molecular Weight: 227.05 g/mol Siedepunkt: 275 °C (2 mmHg) Schmelzpunkt: 194…197 °C |
MDL:
MFCD00001677 CAS-Nummer: 117-80-6 EINECS: 204-210-5 UN: 2811 ADR: 6.1 III Merck Index: 13,03071 |
Spezifikation Testergebnisse
Appearance | Yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 194 to 197 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...