6-Brompiperonal ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 6-Brom-1,3-benzodioxol-5-carbaldehyd

A17510.06 A17510.14 A17510.22
A17510.06EA 59.9 EUR
A17510.06 A17510.14 A17510.22
6-Brompiperonal ≥98%
6-Brompiperonal

Purity: 98% CAS No.: 15930-53-7 Molecular Formula: C8H5BrO3 Formula Weight: 229.03 Melting Point: 127-130° MDL No.: MFCD00022952

Formel: C₈H₅BrO₃
Molecular Weight: 229.03 g/mol
Schmelzpunkt: 127…130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00022952
CAS-Nummer: 15930-53-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR