4-Brom-2,2-difluor-1,3-benzodioxol ≥97%
Lieferant: Thermo Scientific
Achtung
Synonyme: 4-Bromo-2,2-difluorobenzo[d][1,3]dioxole
B24389.03 B24389.06
B24389.03EA 54.8 EUR
B24389.03 B24389.06
4-Brom-2,2-difluor-1,3-benzodioxol ≥97%
4-Brom-2,2-difluor-1,3-benzodioxol
Purity: 97% CAS No.: 144584-66-7 Molecular Formula: C7H3BrF2O2 Formula Weight: 237.00 Refractive Index: 1.4968
Formel: C₇H₃BrF₂O₂
Molecular Weight: 237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631385
CAS-Nummer: 144584-66-7