5-Brom-2,2-difluor-1,3-benzodioxol

Lieferant: CHEMPUR
Achtung

Synonyme: 5-Bromo-2,2-difluorobenzodioxole, 5-bromo-2,2-difluoro-2H-1,3-benzodioxole

FL7832.5 FL7832.25
CHMPFL7832.5EA 0 EUR
CHMPFL7832.5 CHMPFL7832.25
5-Brom-2,2-difluor-1,3-benzodioxol
5-Brom-2,2-difluor-1,3-benzodioxol
Formel: C₇H₃BrF₂O₂
Molecular Weight: 237 g/mol
MDL: MFCD00236212
CAS-Nummer: 33070-32-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR