5-Amino-2,2-difluor-1,3-benzodioxol ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2,2-Difluoro-1,3-benzodioxol-5-amine, 3,4-(Difluoromethylenedioxy)aniline
B24336.03
B24336.06
B24336.03EA
40.7
EUR
B24336.03
B24336.06
5-Amino-2,2-difluor-1,3-benzodioxol ≥97%
5-Amino-2,2-difluor-1,3-benzodioxol
Purity: 97+%
CAS No.: 1544-85-0
Molecular Formula: C7H5F2NO2
Formula Weight: 173.12
Refractive Index: 1.4990
Odour: Recognizable
Boiling Point: 95-97°/12mm
Beilstein Reference: 1343593
UN No.: 2810
MDL No.: MFCD00190144
Formel:
C₇H₅F₂NO₂ Molecular Weight: 173,12 g/mol Siedepunkt: 95…97 °C (12 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00190144 CAS-Nummer: 1544-85-0 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...