1,4-Benzodioxan ≥98%
Lieferant: Thermo Scientific
Achtung
|
Synonyme:
2,3-Dihydrobenzo[b][1,4]dioxine, 2,3-Dihydro-1,4-benzodioxin, Benzo-1,4-dioxane, 1,4-Benzodioxan
A17558.09
A17558.18
A17558.30
A17558.09EA
26.5
EUR
A17558.09
A17558.18
A17558.30
1,4-Benzodioxan ≥98%
1,4-Benzodioxan
Purity: 98%
CAS No.: 493-09-4
Molecular Formula: C8H8O2
Formula Weight: 136.15
Flash Point: 87°(188°F)
Refractive Index: 1.5490
Boiling Point: 216°
Beilstein Reference: 120846
MDL No.: MFCD00006821
Formel:
C₈H₈O₂ Molecular Weight: 136,15 g/mol Siedepunkt: 216 °C Dichte: 1,170 Flammpunkt: 87 °C (188 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006821 CAS-Nummer: 493-09-4 EINECS: 207-775-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...