2-Cyanphenylboronsäure 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Dihydroxy(2-cyanphenyl)boran, 2-Cyanbenzolboronsäure, (2-Cyanphenyl)metaborsäure

365960010 365960050
ACRO365960010EA 59.2 EUR
ACRO365960010 ACRO365960050
2-Cyanphenylboronsäure 98%
2-Cyanphenylboronsäure
Formel: C₇H₆BNO₂
Molecular Weight: 146,94 g/mol
Schmelzpunkt: >350 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01632208
CAS-Nummer: 138642-62-3

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to cream Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR