2-Cyanphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Dihydroxy(2-cyanphenyl)boran, 2-Cyanbenzolboronsäure, (2-Cyanphenyl)metaborsäure

M-Clarity™ Qualitätsstufe = MQ100

521396-1G 521396-5G
SIAL521396-1GEA 49.8 EUR
SIAL521396-1G SIAL521396-5G
2-Cyanphenylboronsäure, Sigma-Aldrich®
2-Cyanphenylboronsäure
Formel: C₇H₆BNO₂
Molecular Weight: 146,94 g/mol
MDL: MFCD01632208
CAS-Nummer: 138642-62-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR