3-Cyanphenylboronsäure ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(3-Cyanophenyl)metaboric acid, Dihydroxy(3-cyanophenyl)borane, 3-Cyanobenzeneboronic acid
L19635.03
L19635.06
L19635.03EA
51.3
EUR
L19635.03
L19635.06
3-Cyanphenylboronsäure ≥98%
3-Cyanphenylboronsäure
Purity: 98+%
CAS No.: 150255-96-2
Molecular Formula: C7H6BNO2
Formula Weight: 146.94
Melting Point: ca 298° dec.
Beilstein Reference: 6136720
Formel:
C₇H₆BNO₂ Molecular Weight: 146,94 g/mol Schmelzpunkt: ∼ 298 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01318967 CAS-Nummer: 150255-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...