4-Cyanphenylboronsäure 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Cyanobenzeneboronic acid, (4-Cyanophenyl)metaboric acid, Dihydroxy(4-cyanophenyl)borane

359040010 359040100
ACRO359040010EA 74.9 EUR
ACRO359040010 ACRO359040100
4-Cyanphenylboronsäure 97%
4-Cyanphenylboronsäure
Formel: C₇H₆BNO₂
Molecular Weight: 146,94 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01318968
CAS-Nummer: 126747-14-6

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR