4-Cyanphenylboronsäure ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 4-Cyanobenzeneboronic acid, (4-Cyanophenyl)metaboric acid, Dihydroxy(4-cyanophenyl)borane

L18007.03 L18007.06
L18007.03EA 104 EUR
L18007.03 L18007.06
4-Cyanphenylboronsäure ≥98%
4-Cyanphenylboronsäure

Purity: 98% CAS No.: 126747-14-6 Molecular Formula: C7H6BNO2 Formula Weight: 146.94 Melting Point: 300° Beilstein Reference: 6593772

Formel: C₇H₆BNO₂
Molecular Weight: 146,94 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318968
CAS-Nummer: 126747-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR