4-Cyanphenylboronsäure
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
4-Cyanobenzeneboronic acid, (4-Cyanophenyl)metaboric acid, Dihydroxy(4-cyanophenyl)borane
FL10788.2
010788-5G
010788-25G
CHMPFL10788.2EA
0
EUR
CHMPFL10788.2
FLUO010788-5G
FLUO010788-25G
4-Cyanphenylboronsäure
4-Cyanphenylboronsäure
Formel:
C₇H₆BNO₂ Molecular Weight: 146,94 g/mol Schmelzpunkt: > 300 °C |
MDL:
MFCD01318968 CAS-Nummer: 126747-14-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...