3-Nitrophenylboronsäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Dihydroxy(3-nitrophenyl)boran, 3-Nitrobenzolboronsäure, (3-Nitrophenyl)metaborsäure

344700010 344700050
ACRO344700010EA 28.7 EUR
ACRO344700010 ACRO344700050
3-Nitrophenylboronsäure 97%
3-Nitrophenylboronsäure
Formel: C₆H₆BNO₄
Molecular Weight: 166.93 g/mol
Schmelzpunkt: 289…291 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00007193
CAS-Nummer: 13331-27-6

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow to light brown Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR