3-Nitrophenylboronsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Dihydroxy(3-nitrophenyl)boran, 3-Nitrobenzolboronsäure, (3-Nitrophenyl)metaborsäure

A13336.03 A13336.06 A13336.14
A13336.03EA 30.7 EUR
A13336.03 A13336.06 A13336.14
3-Nitrophenylboronsäure ≥98%
3-Nitrophenylboronsäure

Purity: 98% CAS No.: 13331-27-6 Molecular Formula: C6H6BNO4 Formula Weight: 166.93 Melting Point: ca 285° dec. Beilstein Reference: 2938638

Formel: C₆H₆BNO₄
Molecular Weight: 166.93 g/mol
Schmelzpunkt: ∼ 285 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007193
CAS-Nummer: 13331-27-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR