4-Nitrophenylboronsäure 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(4-Nitrophenyl)metaborsäure, 4-Nitrobenzolboronsäure, Dihydroxy(4-nitrophenyl)boran
H27767.06
H27767.03
H27767.06EA
201
EUR
H27767.06
H27767.03
4-Nitrophenylboronsäure 95%
4-Nitrophenylboronsäure
Purity: 95%
CAS No.: 24067-17-2
Molecular Formula: C6H6BNO4
Formula Weight: 166.93
Melting Point: ca 285° dec.
MDL No.: MFCD00161360
Formel:
C₆H₆BNO₄ Molecular Weight: 166.93 g/mol Schmelzpunkt: ∼ 285 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00161360 CAS-Nummer: 24067-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...