2-Methoxyphenylboronsäure
Lieferant: CHEMPUR
Synonyme:
(2-Methoxyphenyl)metaborsäure, Dihydroxy(2-methoxyphenyl)boran, 2-Methoxybenzolboronsäure
FL11001.POR
011001-25G
011001-5G
CHMPFL11001.POREA
0
EUR
CHMPFL11001.POR
FLUO011001-25G
FLUO011001-5G
2-Methoxyphenylboronsäure
2-Methoxyphenylboronsäure
Formel:
C₇H₉BO₃ Molecular Weight: 151.96 g/mol Schmelzpunkt: 165 |
MDL:
MFCD00236047 CAS-Nummer: 5720-06-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...