3-Methoxyphenylboronsäure
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
(3-Methoxyphenyl)metaborsäure, Dihydroxy(3-methoxyphenyl)boran, 3-Methoxybenzolboronsäure
FL11002.POR
011002-25G
011002-5G
CHMPFL11002.POREA
0
EUR
CHMPFL11002.POR
FLUO011002-25G
FLUO011002-5G
3-Methoxyphenylboronsäure
3-Methoxyphenylboronsäure
Formel:
C₇H₉BO₃ Molecular Weight: 151.96 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00161359 CAS-Nummer: 10365-98-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...