4-Methoxyphenylboronsäure 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (4-Methoxyphenyl)metaborsäure, Dihydroxy(4-methoxyphenyl)boran, 4-Methoxybenzolboronsäure

309480010 309480050 309480250
ACRO309480010EA 44.6 EUR
ACRO309480010 ACRO309480050 ACRO309480250
4-Methoxyphenylboronsäure 97%
4-Methoxyphenylboronsäure
Formel: C₇H₉BO₃
Molecular Weight: 151,96 g/mol
Schmelzpunkt: 208…210 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039139
CAS-Nummer: 5720-07-0

Bestellen

Spezifikation Testergebnisse

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR