4-Methoxyphenylboronsäure
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
(4-Methoxyphenyl)metaborsäure, Dihydroxy(4-methoxyphenyl)boran, 4-Methoxybenzolboronsäure
003422-5G
003422-25G
FLUO003422-5GEA
0
EUR
FLUO003422-5G
FLUO003422-25G
4-Methoxyphenylboronsäure
4-Methoxyphenylboronsäure
Formel:
C₇H₉BO₃ Schmelzpunkt: 204…207 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039139 CAS-Nummer: 5720-07-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...