2,4-Dimethoxyphenylboronsäure ≥98%

Lieferant: Thermo Scientific
Achtung

B24374.03 B24374.06 B24374.14
B24374.03EA 46.3 EUR
B24374.03 B24374.06 B24374.14
2,4-Dimethoxyphenylboronsäure ≥98%
2,4-Dimethoxyphenylboronsäure

Purity: 98% CAS No.: 133730-34-4 Molecular Formula: C8H11BO4 Formula Weight: 181.98 Melting Point: 115-120° Beilstein Reference: 4744213

Formel: C₈H₁₁BO₄
Molecular Weight: 181,98 g/mol
Schmelzpunkt: 115…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01074590
CAS-Nummer: 133730-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR