2,5-Dimethoxyphenylboronsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (2,5-Dimethoxyphenyl)metaborsäure, 2,5-Dimethoxybenzolboronsäure, Dihydroxy(2,5-dimethoxyphenyl)boran

B24571.03 B24571.06 B24571.14
B24571.03EA 12.8 EUR
B24571.03 B24571.06 B24571.14
2,5-Dimethoxyphenylboronsäure ≥98%
2,5-Dimethoxyphenylboronsäure

Purity: 98% CAS No.: 107099-99-0 Molecular Formula: C8H11BO4 Formula Weight: 181.98 Melting Point: 92-94° Beilstein Reference: 2969872

Formel: C₈H₁₁BO₄
Molecular Weight: 181.98 g/mol
Schmelzpunkt: 92…94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318181
CAS-Nummer: 107099-99-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR