2,6-Dimethoxyphenylboronsäure
Lieferant: FLUOROCHEM
010991-1G
010991-5G
010991-25G
FLUO010991-1GEA
31.1
EUR
FLUO010991-1G
FLUO010991-5G
FLUO010991-25G
2,6-Dimethoxyphenylboronsäure
2,6-Dimethoxyphenylboronsäure
Formel:
C₈H₁₁BO₄ Molecular Weight: 181,98 g/mol |
MDL:
MFCD01318987 CAS-Nummer: 23112-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...