3,4-Dimethoxyphenylboronsäure

Lieferant: FLUOROCHEM

010992-5G 010992-25G
FLUO010992-5GEA 45.9 EUR
FLUO010992-5G FLUO010992-25G
3,4-Dimethoxyphenylboronsäure
3,4-Dimethoxyphenylboronsäure
Formel: C₈H₁₁BO₄
Molecular Weight: 181.98 g/mol
MDL: MFCD01074574
CAS-Nummer: 122775-35-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR