2,3,4-Trimethoxyphenylboronsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(2,3,4-Trimethoxyphenyl)metaborsäure, 2,3,4-Trimethoxybenzolboronsäure, Dihydroxy(2,3,4-trimethoxyphenyl)boran
L19838.03
L19838.06
L19838.03EA
32.7
EUR
L19838.03
L19838.06
2,3,4-Trimethoxyphenylboronsäure ≥98%
2,3,4-Trimethoxyphenylboronsäure
Purity: 98%
CAS No.: 118062-05-8
Molecular Formula: C9H13BO5
Formula Weight: 212.01
Storage Temperature: -30°C to -10°C
Melting Point: 74-79°
Beilstein Reference: 4185924
MDL No.: MFCD01318183
Formel:
C₉H₁₃BO₅ Molecular Weight: 212,01 g/mol Schmelzpunkt: 74…79 °C |
MDL:
MFCD01318183 CAS-Nummer: 118062-05-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...