2,3,4-Trimethoxyphenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: (2,3,4-Trimethoxyphenyl)metaborsäure, 2,3,4-Trimethoxybenzolboronsäure, Dihydroxy(2,3,4-trimethoxyphenyl)boran

011064-1G 011064-5G
FLUO011064-1GEA 36.9 EUR
FLUO011064-1G FLUO011064-5G
2,3,4-Trimethoxyphenylboronsäure
2,3,4-Trimethoxyphenylboronsäure
Formel: C₉H₁₃BO₅
Molecular Weight: 212.01 g/mol
MDL: MFCD01318183
CAS-Nummer: 118062-05-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR