3-Hydroxyphenylboronsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Hydroxybenzolboronsäure, (3-Hydroxyphenyl)metaborsäure, Dihydroxy(3-hydroxyphenyl)boran
L19061.03
L19061.06
L19061.03EA
53.7
EUR
L19061.03
L19061.06
3-Hydroxyphenylboronsäure ≥97%
3-Hydroxyphenylboronsäure
Purity: 97%
CAS No.: 87199-18-6
Molecular Formula: C6H7BO3
Formula Weight: 137.93
Melting Point: 208-210°
Beilstein Reference: 3240771
Formel:
C₆H₇BO₃ Molecular Weight: 137.93 g/mol Schmelzpunkt: 208…210 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01074603 CAS-Nummer: 87199-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...