4-Hydroxyphenylboronsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(4-Hydroxyphenyl)metaboric acid, Dihydroxy(4-hydroxyphenyl)borane, 4-Hydroxybenzeneboronic acid
L15594.03
L15594.06
L15594.03EA
56.8
EUR
L15594.03
L15594.06
4-Hydroxyphenylboronsäure ≥97%
4-Hydroxyphenylboronsäure
Purity: 97%
CAS No.: 71597-85-8
Molecular Formula: C6H7BO3
Formula Weight: 137.93
Melting Point: ca 285° dec.
Beilstein Reference: 2963491
Formel:
C₆H₇BO₃ Molecular Weight: 137.93 g/mol Schmelzpunkt: ∼ 285 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01074628 CAS-Nummer: 71597-85-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...